missing translation for 'onlineSavingsMsg'
Learn More

RIPX Antibody, Novus Biologicals™

Product Code. p-200057570 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18624957 25 μL 25µL
18614428 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18624957 Supplier Novus Biologicals Supplier No. NBP24855825ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RIPX Polyclonal antibody specifically detects RIPX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPX
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias KIAA0871SINGAR1, protein RUFY3, rap2 interacting protein x, Rap2-interacting protein x, RIPx, RUN and FYVE domain containing 3, singar, Singar1, Single axon-regulated protein, single axon-related 1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MSALTPPTDMPTPTTDKITQAAMETIYLCKFRVSMDGEWLCLRELDDISLTPDPEPTHEDPNYLMANERMNLMNMAKLSIKGL
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22902
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.