missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Polymerase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41224
This item is not returnable.
View return policy
Description
Rift Valley Fever Virus Polymerase Polyclonal specifically detects Rift Valley Fever Virus Polymerase in Virus samples. It is validated for ELISA.
Specifications
| Rift Valley Fever Virus Polymerase | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| 230 kDa | |
| 0.1 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| EC=2.7.7.48, Replicase, RVFV_sL_gp1, Transcriptase | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase. The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase. Amino Acid Squence: VKDRPDERNRYHRVHL | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction