missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RICTOR Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148876
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifications
| RICTOR | |
| Polyclonal | |
| Unconjugated | |
| Rictor | |
| 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 253260 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6R327 | |
| Rictor | |
| E. coli-derived recombinant human Rictor, Met1-Leu250. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction