missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal protein L4 Rabbit anti-Human, Clone: 3K6, Proteintech
Rabbit Recombinant Antibody
149.00€ - 439.00€
Specifications
| Antigen | Ribosomal protein L4 |
|---|---|
| Clone | 3K6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17629816
|
Proteintech
80959-1-RR-20UL |
20 μL |
149.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17619816
|
Proteintech
80959-1-RR-100UL |
100 μL |
439.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribosomal protein L4 antibody apecifically detetects Ribosomal protein L4 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
| Ribosomal protein L4 | |
| 1000 μg/mL | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 6124 | |
| Ribosomal protein L4 fusion protein Ag28728 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3K6 | |
| Western Blot, ELISA | |
| Liquid | |
| Human | |
| BC009888 | |
| RPL4 | |
| IgG | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title