missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoJ Antibody (OTI1D7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73892F
This item is not returnable.
View return policy
Description
RhoJ Monoclonal specifically detects RhoJ in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RhoJ | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| RHOJ | |
| Full length recombinant protein of human RhoJ (NP_065714) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D7 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ARHJTC10B, FLJ14445, ras homolog gene family, member J, RASL7B, Ras-like protein family member 7B, RAS-like, family 7, member B, RHOI, rho-related GTP-binding protein RhoJ, Tc10-like GTP-binding protein, TC10-like Rho GTPase, TCLMGC34777 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 57381.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction