missing translation for 'onlineSavingsMsg'
Learn More

RhoG Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen RhoG
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18239775
View Documents
Novus Biologicals
NBP1-79794
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RhoG Polyclonal specifically detects RhoG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RhoG
Unconjugated
RUO
NP_001656
391
Synthetic peptide directed towards the C terminal of human RHOGThe immunogen for this antibody is RHOG. Peptide sequence LVGTKKDLRAQPDTLRRLKEQGQAPITPQQGQALAKQIHAVRYLECSALQ.
Primary
Polyclonal
Rabbit
Cell Cycle and Replication
ARHGMGC125835, MGC125836, ras homolog gene family, member G (rho G), RhoG, rho-related GTP-binding protein RhoG
RHOG
IgG
21 kDa
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.