missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35363-20ul
This item is not returnable.
View return policy
Description
RhoF Polyclonal antibody specifically detects RhoF in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| RhoF | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human RhoF (NP_061907.2).,, Sequence:, SQGSFPEHYAPSVFEKYTASVTVGSKEVTLNLYDTAGQEDYDRLRPLSYQNTHLVLICYDVMNPTSYDNVLIKWFPEVTHF | |
| 20 μL | |
| Signal Transduction | |
| 54509 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction