missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RHOC (Human) Recombinant Protein
Human RHOC full-length ORF ( AAH07245, 1 a.a. - 193 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000389-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAAIRKKLVIVGDGACGKTCLLIVFSKDQFPEVYVPTVFENYIADIEVDGKQVELALWDTAGQEDYDRLRPLSYPDTDVILMCFSIDSPDSLENIPEKWTPEVKHFCPNVPIILVGNKKDLRQDEHTRRELAKMKQEPVRSEEGRDMANRISAFGYLECSAKTKEGVREVFEMATRAGLQVRKNKRRRGCPIL
Specifications
AAH07245 | |
ras homolog gene family, member C | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RHOC | |
GST |
389 | |
Wheat germ expression system | |
10 μg | |
ARH9, ARHC, H9, MGC1448, MGC61427, RHOH9 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |