missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RHOC (Human) Recombinant Protein
Human RHOC full-length ORF ( AAH07245, 1 a.a. - 193 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH07245 |
---|---|
Gene ID (Entrez) | 389 |
Name | ras homolog gene family, member C |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16161831
|
Abnova™
H00000389-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16171831
|
Abnova™
H00000389-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Sequence: MAAIRKKLVIVGDGACGKTCLLIVFSKDQFPEVYVPTVFENYIADIEVDGKQVELALWDTAGQEDYDRLRPLSYPDTDVILMCFSIDSPDSLENIPEKWTPEVKHFCPNVPIILVGNKKDLRQDEHTRRELAKMKQEPVRSEEGRDMANRISAFGYLECSAKTKEGVREVFEMATRAGLQVRKNKRRRGCPIL
Specifications
AAH07245 | |
ras homolog gene family, member C | |
125% SDS-PAGE Stained with Coomassie Blue | |
ARH9, ARHC, H9, MGC1448, MGC61427, RHOH9 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
389 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RHOC | |
GST |