Learn More
Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals
Product Code. p-220857 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 10052590
Brand: Thermo Scientific Acros 196221000
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Rhenium standard solution
Packaging Plastic bottle
CAS 13598-65-7, 7732-18-5
Physical Form Liquid
Molecular Formula H4NO4Re
Concentration 997 to 1003 μg/mL of Re (4+) in water (20°C)
MDL Number MFCD00012377
Color Colorless
Solubility Miscible with water
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
SMILES N.O[Re](=O)(=O)=O
IUPAC Name rheniumoylol amine
Molecular Weight (g/mol) 268.24
PubChem CID 3084163
Quantity 100 mL
Grade AAS
Show More Show Less
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.