Learn More
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Product Code. 10669865 Shop All Thermo Scientific Products
Click to view available options
Type:
Fitting Set, Short
Fitting Set, Standard Length
Fitting Set, X-Long
Replacement Ferrule
Unit Size:
Pack of 5
Product Code. 10669865
Brand: Thermo Scientific™ 6000055
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
TRUSTED_SUSTAINABILITY
Specifications
Type Fitting Set, Short
Description 1/16 in. Short fitting set; 5/Pk.
For Use With (Equipment) PEEK Tubing
Material PEEK