missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHBDL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
391.65€
Specifications
| Antigen | RHBDL2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RHBDL2 Polyclonal specifically detects RHBDL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RHBDL2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 54933 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YAVWKPQKQWITLDTGILESPFIYSPEKREEAWRFISY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 3.4.21.105, FLJ20435, MGC16997, rhomboid (veinlet, Drosophila)-like 2, rhomboid protease 2, rhomboid, veinlet-like 2 (Drosophila), Rhomboid-like protein 2, rhomboid-related protein 2, RRP2 | |
| RHBDL2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title