missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The RGS5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RGS5. This antibody recognizes human, mouse, rat antigen. The RGS5 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | RGS5 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, RGS5 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 8490 |
| Target Species | Human, Mouse, Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?