missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody (CL5568) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61419V
This item is not returnable.
View return policy
Description
RGS5 Monoclonal antibody specifically detects RGS5 in Human samples. It is validated for Western Blot
Specifications
| RGS5 | |
| Monoclonal | |
| DyLight 405 | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| This antibody was developed using a synthetic peptide derived from O15539, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 8490 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL5568 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction