missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94534-0.1ml
This item is not returnable.
View return policy
Description
RGS13 Polyclonal antibody specifically detects RGS13 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| RGS13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC17173, regulator of G-protein signaling 13, regulator of G-protein signalling 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-159 of human RGS13 (NP_002918.1). MSRRNCWICKMCRDESKRPPSNLTLEEVLQWAQSFENLMATKYGPVVYAAYLKMEHSDENIQFWMACETYKKIASRWSRISRAKKLYKIYIQPQSPREINIDSSTRETIIRNIQEPTETCFEEAQKIVYMHMERDSYPRFLKSEMYQKLLKTMQSNNSF | |
| 0.1 mL | |
| Signal Transduction | |
| 6003 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction