missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGM-A Antibody (CL11342) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08000MFV610
This item is not returnable.
View return policy
Description
RGM-A Monoclonal antibody specifically detects RGM-A in Human samples. It is validated for Western Blot
Specifications
| RGM-A | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A | |
| This antibody was developed using a synthetic peptide derived from Q96B86, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience, Stem Cells | |
| 56963 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot | |
| CL11342 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction