missing translation for 'onlineSavingsMsg'
Learn More

Abcam RGD, Cell Adhesion Peptide, >95% Purity, Solid, 5 mg

Product Code. 32109041 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109041 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109041 Supplier Abcam Supplier No. ab1426985mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell adhesion peptide sequence with >95% purity.

  • Molecular Weight: 346.34 Da
  • Purity: >95%
  • Form: Solid
  • Composition: 100% RGD
  • Citations: 8

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Peptide-based Substrate
Target Molecule RGD (Arg-Gly-Asp)
Formulation 100% RGD
Quantity 5 mg
For Use With (Application) Cell adhesion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.