missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFXANK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97640-50ul
187.85 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
RFXANK Polyclonal antibody specifically detects RFXANK in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| RFXANK | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 μL/mg of lysate | |
| ANKRA1Regulatory factor X subunit B, ankyrin repeat-containing regulatory factor X-associated protein, BLS, DNA-binding protein RFXANK, F14150_1, MGC138628, regulatory factor X-associated ankyrin-containing proteinAnkyrin repeat family A protein 1, RFXB, RFX-BRFX-Bdelta4 | |
| Produced in rabbits immunized with E. coli-derived Human RFXANK fragment. | |
| 50 μg | |
| Adaptive Immunity, Immunology | |
| 8625 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction