Learn More
Brady™ Retro Reflective All-weather Labels
Product Code. 17931960
Click to view available options
Dimensions (LxW):
12.70 mm (W) x 6.10 m (L)
19.05 mm (W) x 6.10 m (L)
25.40 mm (W) x 6.10 m (L)
38.10 mm (W) x 6.10 m (L)
Color:
White
Yellow
For Use With (Equipment):
BMP41, BMP51, BMP53, M511, M410, M510
BMP51, BMP53, M511, M510
Unit Size:
Each
Product Code. 17931960
Brand: Brady™ 170812
Only null left
Add to basket
Add to basket
Increase safety and visibility with the B-584 highly-reflective label material. Print labels almost anywhere high visibility is needed with a compact Brady™ handheld label printer.
  • Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required
  • Resistant to abrasions, weather, solvents and extreme temperatures
  • Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions
  • Able to withstand outdoor conditions for 4 to 6 years on average
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance 2011/65/EU, 2015/863
Adhesive Type Permanent Acrylic
Coating None
Color Yellow
Finish Gloss
Format Continuous
Material Plastic
Printable Sides Top
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling;Facility identification;General identification;Safety identification
For Use With (Equipment) BMP41, BMP51, BMP53, M511, M410, M510
Min. Operating Temperature (Metric) -40°C
Model M4C-750-584-YL
No. per Pack 1 Label(s) / Box
Quantity 1
Size 19.05 mm (W) x 6.10 m (L)
Temperature Range (English) -40°F to 158°F
Temperature Range (Metric) -40°C to 70°C
Thickness (English) 0.0055 in
Thickness (Metric) 0.14 mm
Dimensions (LxW) 19.05 mm (W) x 6.10 m (L)
Length (English) 20 ft.
Length (Metric) 6.10 m
Width (English) 0.75 in
Width (Metric) 19.05 mm
Width (English) Printable Area 19.05 mm
Height (English) Printable Area 0.5 in.
Alternate Part Number M4C-750-584-YL