Learn More
Brady™ Retro Reflective All-weather Labels
Increase safety and visibility with the B-584 highly-reflective label material. Print labels almost anywhere high visibility is needed with a compact Brady™ handheld label printer.
42.05€ - 66.54€
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Certifications/Compliance | 2011/65/EU, 2015/863 |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Finish | Gloss |
| Product Code | Brand | Dimensions (L x W) | Color | For Use With (Equipment) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Dimensions (L x W) | Color | For Use With (Equipment) | Price | Quantity & Availability | |||
|
17981940
|
Brady™
170809 |
12.70 mm (W) x 6.10 m (L) | White | BMP41, BMP51, BMP53, M511, M410, M510 |
42.05€
Pack of 1 |
In Stock
Log in to see stock. |
|||
|
17971950
|
Brady™
170810 |
12.70 mm (W) x 6.10 m (L) | Yellow | BMP41, BMP51, BMP53, M511, M410, M510 |
42.05€
Pack of 1 |
In Stock
Log in to see stock. |
|||
|
17971930
|
Brady™
170811 |
19.05 mm (W) x 6.10 m (L) | White | BMP41, BMP51, BMP53, M511, M410, M510 |
49.00€
Pack of 1 |
In Stock
Log in to see stock. |
|||
|
17931960
|
Brady™
170812 |
19.05 mm (W) x 6.10 m (L) | Yellow | BMP41, BMP51, BMP53, M511, M410, M510 |
49.00€
Pack of 1 |
In Stock
Log in to see stock. |
|||
|
17801769
|
Brady™
170935 |
25.40 mm (W) x 6.10 m (L) | White | BMP51, BMP53, M511, M510 |
55.90€ 53.30€ / Pack of 1 Save 2.60€ 4% Off |
Estimated Shipment: 25-11-2026
Log in to see stock. |
|||
|
17831759
|
Brady™
170936 |
25.40 mm (W) x 6.10 m (L) | Yellow | BMP51, BMP53, M511, M510 |
55.90€ 53.30€ / Pack of 1 Save 2.60€ 4% Off |
In Stock
Log in to see stock. |
|||
|
17861739
|
Brady™
170937 |
38.10 mm (W) x 6.10 m (L) | White | BMP51, BMP53, M511, M510 |
69.80€ 66.54€ / Pack of 1 Save 3.26€ 4% Off |
In Stock
Log in to see stock. |
|||
|
17851739
|
Brady™
170938 |
38.10 mm (W) x 6.10 m (L) | Yellow | BMP51, BMP53, M511, M510 |
69.80€ 66.54€ / Pack of 1 Save 3.26€ 4% Off |
In Stock
Log in to see stock. |
|||
Description
- Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required
- Resistant to abrasions, weather, solvents and extreme temperatures
- Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions
- Able to withstand outdoor conditions for 4 to 6 years on average
Specifications
| Not Autoclavable | |
| Permanent Acrylic | |
| Gloss | |
| Plastic | |
| Non-sterile | |
| -40°C | |
| 1 | |
| -40°C to 70°C | |
| 0.14 mm | |
| 6.10 m |
| 2011/65/EU, 2015/863 | |
| None | |
| Continuous | |
| Thermal Transfer | |
| Component and equipment labelling;Facility identification;General identification;Safety identification | |
| 1 Label(s) / Box | |
| -40°F to 158°F | |
| 0.0055 in | |
| 20 ft. | |
| 0.5 in. |