missing translation for 'onlineSavingsMsg'
Learn More

Retinol Binding Protein RBP Mouse anti-Human, Mouse, Rat, Goat, Chimpanzee, Monkey, Rabbit, Clone: rRBP1/872, Novus Biologicals™

Mouse Monoclonal Antibody

499.00€

Specifications

Antigen Retinol Binding Protein RBP
Clone rRBP1/872
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18675196
View Documents
Novus Biologicals
NBP3-07197-100ug
100 μg
499.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Retinol Binding Protein RBP Monoclonal specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat, Goat, Chimpanzee, Monkey, Rabbit samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Retinol Binding Protein RBP
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human, Mouse, Rat, Goat, Chimpanzee
C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular
Recombinant full-length human Retinol Binding Protein RBP protein
Primary
Store at 4°C.
rRBP1/872
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
5947
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.