missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (342) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43763JF669
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for ELISA,Lateral Flow
Specifications
| Respiratory Syncytial Virus Glycoprotein F | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Lateral Flow | |
| 342 | |
| ELISA, Lateral Flow Assay | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (Uniprot#: P03420-1; Met 1-Thr 529). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction