missing translation for 'onlineSavingsMsg'
Learn More

Abcam Resorufin Butyrate, Fluorogenic Lipase Substrate, 25 mg

Product Code. 32117233
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117233 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117233 Supplier Abcam Supplier No. ab14543525mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorogenic substrate for the detection of lipase and cholinesterase activity.

  • Molecular Weight: 283.28 Da
  • Purity: >95%
  • Application: Fluorogenic lipase and cholinesterase substrate; fluoresces on hydrolysis
  • Analog: Resorufin methyl ether (ab145372) analog

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Resorufin butyrate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Resorufin butyrate
Target Molecule Lipase, Cholinesterase
Detection Method Fluorescence
Formulation 100% Resorufin butyrate
Quantity 25 mg
For Use With (Application) Lipase assay, cholinesterase assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.