missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcroBiosystems resDetectTM Cas9 (CRISPR Associated Protein 9) ELISA Kit (Residue Testing)
Description
resDetectTM Cas9 (CRISPR Associated Protein 9) ELISA Kit (Residue Testing) is expressed from Streptococcus pyogenes
Specifications
Specifications
| Format | Solid |
| Quantity | 96 Tests |
| Species | S. pyogenes |
| Content And Storage | 1. Unopened kit should be stored at 2°C to 8°C upon receiving. 2. Find the expiration date on the outside packaging and do not use reagents past their expiration date. 3. The opened kit should be stored per components table. The shelf life is 30 days from the date of opening. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?