Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules 
Product Code. p-7114366
Click to view available options
For Use With (Equipment):
For 0.1 to 0.25 mm Columns
For 1/32 in. Columns
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 12937111
Brand: Trajan Scientific and Medical™ 073470
Only null left
Add to basket
Add to basket
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Type SilTite Mini Union Ferrules for 0.1 to 0.25 mm ID Columns
Diameter (Metric) Inner Tubing 0.1 to 0.25 mm
For Use With (Equipment) For 0.1 to 0.25 mm Columns
Quantity 10/Pk.