missing translation for 'onlineSavingsMsg'
Learn More

Repaglinide Methyl Ester, Mikromol™

Product Code. 30049752
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049752

Brand: Mikromol MM0944.070025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Repaglinide Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Repaglinide Methyl Ester
CAS 800365-89-3
Analyte or Component Names Repaglinide Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C28 H38 N2 O4
Certifications/Compliance ISO 9001
Synonym Repaglinide Methyl Ester, 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]-benzoic acid methyl ester, (S)-2-Ethoxy-4-[[[N-[1-(2-piperidinophenyl)-3-methylbutyl]amino]carbonyl]methyl]benzoic acid methyl ester
InChI Formula InChI=1S/C28H38N2O4/c1-5-34-26-18-21(13-14-23(26)28(32)33-4)19-27(31)29-24(17-20(2)3)22-11-7-8-12-25(22)30-15-9-6-10-16-30/h7-8,11-14,18,20,24H,5-6,9-10,15-17,19H2,1-4H3,(H,29,31)/t24-/m0/s1
SMILES CCOc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)ccc1C(=O)OC
IUPAC Name methyl 2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoate
Molecular Weight (g/mol) 466.61
Quantity 25 mg
Formula Weight 466.2832 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.