Learn More
Novus Biologicals™ RelA/NFkB p65 [p Ser529, p Ser536] Inhibitor Peptide Set
Product Code. p-5538755 Shop All Bio Techne Products
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Product Code. 18789733
Brand: Novus Biologicals™ NBP229321
For use in research applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 2 mg