Learn More
Novus Biologicals™ RelA/NFkB p65 [p Ser529, p Ser536] Inhibitor Peptide Set
Product Code. 18789733 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789733 2 mg 2mg
18780004 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18789733 Supplier Novus Biologicals™ Supplier No. NBP229321
  • ONLINE EXCLUSIVE PRICE
    842.00€
    795.90€ / 2mg
    Save 46.10€
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
For use in research applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 2 mg