missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg3G Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 470.00€
Specifications
| Antigen | Reg3G |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:100 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694991
|
Novus Biologicals
NBP2-93737-0.02ml |
0.02 mL |
204.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634261
|
Novus Biologicals
NBP2-93737-0.1ml |
0.1 mL |
470.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reg3G Polyclonal antibody specifically detects Reg3G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Reg3G | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| LPPM429, MGC118999, MGC119001, Pancreatitis-associated protein 1B, Pancreatitis-associated protein IB, PAP IB, PAP-1B, PAP1BMGC118998, PAPIB, Reg III-gamma, REG-3-gamma, regenerating gene III, regenerating islet-derived 3 gamma, regenerating islet-derived protein 3-gamma, Regenerating islet-derived protein III-gamma, REG-III, UNQ429 | |
| A synthetic peptide corresponding to a sequence within amino acids 27-174 of mouse Reg3G (NP_035390.1). EVAKKDAPSSRSSCPKGSRAYGSYCYALFSVSKNWYDADMACQKRPSGHLVSVLSGAEASFLSSMIKSSGNSGQYVWIGLHDPTLGYEPNRGGWEWSNADVMNYINWETNPSSSSGNHCGTLSRASGFLKWRENYCNLELPYVCKFKA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 130120 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title