missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG2 Monoclonal Antibody (239237)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561099
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| REG2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q08731 | |
| Reg2 | |
| E. coli-derived recombinant mouse Reg2. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 239237 | |
| Unconjugated | |
| Reg2 | |
| islet of Langerhans regenerating protein 2; lithostathine 1 beta; lithostathine-1-beta; Lithostathine-2; pancreatic stone protein 2; Pancreatic thread protein 2; PSP; PSP-2; PSPS2; PTP; REG1B; REG-1-beta; Reg2; REG-2; regenerating family member 1 beta; regenerating islet-derived 1 beta (pancreatic stone protein, pancreatic thread protein); regenerating islet-derived 2; regenerating islet-derived protein 1-beta; regenerating islet-derived, mouse homolog 2; regenerating protein I beta; REGH; REGI-BETA; RegII; REGL; secretory pancreatic stone protein 2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 19693 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction