missing translation for 'onlineSavingsMsg'
Learn More

Refractive Index Certified Reference Material at 20 °C, 25 °C and 30 °C (Nominal RI Value 1.4196 at 25 °C), UKAS ISO 17025 / ISO 17034 Certified, in Tamper-Evident Glass Vials, 10 mL, Paragon Scientific™

Product Code. 16050290
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 16050290

Brand: Paragon Scientific PSRI04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon Scientific’s range of Refractive Index Certified Reference Materials (CRMs) provide certified values for refractive index measurements at 20 °C, 25 °C and 30 °C and cover a wide range of calibration needs.

  • Refractive Index Certified Reference Materials at 20 °C, 25 °C and 30 °C
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • All measurements are fully traceable to NIST and international protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data at hand and dependable results
  • Available in single 10 mL volume tamper-evident glass bottles or as a multi-pack of 5 per material (includes set of disposable pipettes)
  • 12 month shelf life
  • Manufactured in the United Kingdom

Specifications

Quantity 10 mL
compliance-icons compliance-icons
Product Identifier
  • Refractive Index Standard at 20 °C 25 °C and 30 °C
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : n-dodecane

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.