Learn More
Eppendorf Reference™ 2, Mechanical Single-Channel Pipettes with Fixed Volume
Product Code. 15793509 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
1 μL
2 μL
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
2 mL
2.5 mL
Accuracy:
0.6
0.7
0.8
1.0
1.5
2.0
2.5
Color:
Blue
Dark Gray
Gray
Light Gray
Medium Gray
Red
Yellow
Precision:
0.012
0.015
0.025
0.075
0.2
0.3
0.35
0.4
0.6
1.2
1.5
3.0
6.0
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Accuracy Color Precision Unit Size
15793509 200 μL 0.6 Blue 1.2 Each
15793499 2 μL 2.0 Dark Gray 0.4 Each
15713519 500 μL 0.6 Blue 3.0 Each
15753509 25 μL 0.8 Yellow 0.2 Each
15783499 1 μL 2.5 Dark Gray 0.025 Each
15723519 1000 μL 0.6 Blue 6.0 Each
15783509 200 μL 0.6 Yellow 1.2 Each
15763509 50 μL 0.7 Yellow 0.35 Each
15703509 5 μL 1.5 Medium Gray 0.075 Each
15743519 2.5 mL 0.6 Red 0.015 Each
15703519 250 μL 0.6 Blue 1.5 Each
15733519 2 mL 0.6 Red 0.012 Each
15713509 10 μL 1.0 Gray 0.3 Each
15733509 20 μL 0.8 Light Gray 0.3 Each
15723509 10 μL 1.0 Yellow 0.3 Each
15773509 100 μL 0.6 Yellow 0.6 Each
15743509 20 μL 0.8 Yellow 0.3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15793509 Supplier Eppendorf Supplier No. 4925000120
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
Bulk Quote Request
Proven premium precision and accuracy, a long service life, and an ergonomic design
Provides the most accurate results possible alongside robust, reliable handling and optimum user safety. Ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. Optimal choice when quality and reproducibility are essential.
Well Designed Pipet
  • One button operating system lets users quickly move through their pipet tasks; users do not have to move the thumb from side to side to eject tips
  • Ergonomic design for lower operating forces for aspiration/dispensing/tip ejection; limits strain on the user
  • Spring loaded tip cone allows simple, quick tip ejection with active aerosol reduction; improves safety and user reproducibility
  • Color coded/ volume labeled for quick identification of the volume size/ tip size
  • High precision and accuracy provides reliable pipetting results
  • Customizable labeling area
Accurate Settings
  • Volume lock: a quick and secure setting prevents accidental changes and with an always visible 4 digit display, provides increased security is provided for the most stringent lab environments
  • Secondary user adjustment offers the most accurate transfer of liquids different than water without the need for a full calibration
  • 4-digit display shows the exact volume setting and is clearly visible from every angle
  • Ergonomic Design
  • Round upper part makes it possible to work in every position
  • To avoid accidental tip ejections, clear haptic feedback is felt after the blowout step
Lower operating forces for aspiration/dispensing/tip ejection; limits strain on the user
Well Built Instrument
  • Unique surface: few rough edges, smooth surface for comfortable working and simple cleaning
  • Sturdy upper handle guarantees long service life and increased robustness
  • Movable parts and external edges made from stainless steel equip the pipette with outstanding robustness at potential impact sites and for internal components
  • The quick connect lower part enables easy cleaning and maintenance of your pipet
  • Fully autoclavable
Heightened Traceability
  • Serial number is printed on multiple components of the pipette
  • Internal RFID technology: contains all relevant pipette data and TrackIT software from Eppendorf for simple identification and documentation of each pipette in the lab
TRUSTED_SUSTAINABILITY
Specifications
Accuracy 0.6
Autoclavable Autoclavable
Color Blue
For Use With Pipette Tip
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 200 μL
Precision 1.2