Description
- Rectangular Centrifuge Buckets
- Compatible with Eppendorf rotors
- Available in packs of two and four
Specifications
Specifications
| For Use With (Equipment) | Eppendorf rotor A-4-81 (CFA-152-020K) |
| Capacity | 400 mL |
| Container Type | Bucket |
| Quantity | 4/Pk. |