missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish SOD1 / Superoxide Dismutase Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6714-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_571369.1 | |
SOD1 / Superoxide Dismutase Protein | |
Research Use Only | |
sod1 | |
Recombinant Protein | |
E. coli | |
MVNKAVCVLK GTGEVTGTVY FNQEGEKKPV KVTGEITGLT PGKHGFHVHA FGDNTNGCIS AGPHFNPHDK THGGPTDSVR HVGDLGNVTA DASGVAKIEI EDAMLTLSGQ HSIIGRTMVI HEKEDDLGKG GNEESLKTGN AGGRLACGVI GITQ | |
Greater than 86.2% as determined by SDS-PAGE. |
30553 | |
100 μg | |
Not determined. | |
Full Length | |
Zebrafish | |
His-SUMO | |
Tris-based buffer, 50% glycerol |