missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish SOD1 / Superoxide Dismutase Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_571369.1 |
---|---|
Gene ID (Entrez) | 30553 |
Name | SOD1 / Superoxide Dismutase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015320
|
enQuireBio™
QP6714-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16035320
|
enQuireBio™
QP6714-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16025320
|
enQuireBio™
QP6714-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16045320
|
enQuireBio™
QP6714-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055320
|
enQuireBio™
QP6714-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065320
|
enQuireBio™
QP6714-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_571369.1 | |
SOD1 / Superoxide Dismutase Protein | |
Not determined. | |
Full Length | |
Zebrafish | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
30553 | |
Research Use Only | |
sod1 | |
Recombinant Protein | |
E. coli | |
MVNKAVCVLK GTGEVTGTVY FNQEGEKKPV KVTGEITGLT PGKHGFHVHA FGDNTNGCIS AGPHFNPHDK THGGPTDSVR HVGDLGNVTA DASGVAKIEI EDAMLTLSGQ HSIIGRTMVI HEKEDDLGKG GNEESLKTGN AGGRLACGVI GITQ | |
Greater than 86.2% as determined by SDS-PAGE. |