Learn More
enQuireBio™ Recombinant Xenopus Protein Wnt-8 Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) Protein Wnt-8, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_001081637.1 |
---|---|
Gene ID (Entrez) | 397970 |
Name | Protein Wnt-8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088501
|
enQuireBio™
QP9601-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16028511
|
enQuireBio™
QP9601-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16098501
|
enQuireBio™
QP9601-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16018511
|
enQuireBio™
QP9601-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038511
|
enQuireBio™
QP9601-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16008511
|
enQuireBio™
QP9601-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001081637.1 | |
Protein Wnt-8 Protein | |
Not determined. | |
Full Length | |
Xenopus | |
His | |
Tris-based buffer, 50% glycerol |
397970 | |
Research Use Only | |
wnt8 | |
Recombinant Protein | |
Yeast | |
AWSVNNFLMT GPKAYLTYSA SVAVGAQNGI EECKYQFAWE RWNCPESTLQ LATHNGLRSA TRETSFVHAI SSAGVMYTLT RNCSMGDFDN CGCDDSRNGR IGGRGWVWGG CSDNAEFGER ISKLFVDGLE TGQDARALMN LHNNEAGRLA VKETMKRTCK CHGISGSCSI QTCWLQLAEF RDIGNHLKIK HDQALKLEMD KRKMRSGNSA DNRGAIADAF SSVAGSELIF LEDSPDYCLK NISLGLQGTE GRECLQSGKN LSQWERRSCK RLCTDCGLRV EEKKTEIISS CNCKFHWCCT VKCEQCKQVV IKHFCARRER DSNMLNTKRK NRGHRR | |
Greater than 90% as determined by SDS-PAGE. |