missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Recombinant Virus Dengue Virus 3 VLP Sri Lanka D3/H/IMTSSA-SRI/2000/1266 Protein

Product Code. p-7229492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371135 0.1 mg 0.10mg
18355234 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18371135 Supplier Novus Biologicals Supplier No. NBP3148140.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) SDS-PAGE
Formulation 10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias polyprotein
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Dengue Virus 3 VLP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.