missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral B18R Protein
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
The Recombinant Viral B18R Protein is derived from HEK293. The Recombinant Viral B18R Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3707577 |
| Molecular Weight (g/mol) | 65 kDa (monomer) |
| Name | B19R |
| Quantity | 25 μg |
| Source | Human embryonic kidney cell,HEK293-derived viral B18R protein, (N-terminus) MD Human IgG1 (Pro100-Lys300) IEGR Viral B18R (His20-Glu351) Accession &Num; P25213 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction