Learn More
Recombinant VHH Alpaca, anti-Rabbit, Unconjugated, Clone: CTK0102, Proteintech ChromoTek
Product Code. 17323503
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17323503 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17323503 Supplier Chromotek Supplier No. srbGCys22500
 Restricted Item
In Stock
Add to basket
Add to basket
Alpaca Monoclonal Antibody
500 μg Nano-Secondary™ for rabbit IgG: alpaca monoclonal Nanobody, Fab-specific VHH, for 2x Cys conjugation, clone [CTK0102].
Specifications
Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Clone CTK0102
Host Species Alpaca
Quantity 500 μg
Target Species Rabbit