missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant S. cerevisiae ATP Sulfurylase/MET3 Protein

Product Code. 18301688
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 18301688

Brand: R&D Systems™ 7175AS020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to sulfurylate ATP. The specific activity is >2,500 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08536
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol and DTT.
Gene ID (Entrez) 853466
Molecular Weight (g/mol) M.W. (Observed): 57-58 kDa, reducing conditions; M.W. (theoretical): 59 kDa
Name ATP-Sulfurylase/MET3
Quantity 20 μg
Source E. coli-derived yeast ATP-Sulfurylase/MET3 protein Pro2-Phe511, with N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol MET3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ATP-Sulfurylase/MET3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.