missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Staphopain B Protein

A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8022-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032211

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

WP_001088793.1
200 μg
Not determined.
Full Length
S. aureus
GST
Tris-based buffer, 50% glycerol
S. aureus Staphopain B Protein
Research Use Only
sspB
Recombinant Protein
E. coli
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCATFPNQM IEYGKSQGRD IHYQEGVPSY NQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers