missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Staphopain B Protein
A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | WP_001088793.1 |
---|---|
Name | S. aureus Staphopain B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | sspB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002211
|
enQuireBio™
QP8022-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16022211
|
enQuireBio™
QP8022-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16012211
|
enQuireBio™
QP8022-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16032211
|
enQuireBio™
QP8022-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042211
|
enQuireBio™
QP8022-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052211
|
enQuireBio™
QP8022-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
WP_001088793.1 | |
Research Use Only | |
sspB | |
Recombinant Protein | |
E. coli | |
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCATFPNQM IEYGKSQGRD IHYQEGVPSY NQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Staphopain B Protein | |
Not determined. | |
Full Length | |
S. aureus | |
GST | |
Tris-based buffer, 50% glycerol |