missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Enolase Protein

Product Code. p-7153970
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16007880

Brand: enQuireBio™ QP7255ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Enolase, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001121760.1
Gene ID (Entrez) 31213594
Name S. aureus Enolase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol eno
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag His-SUMO
Sequence MPIITDVYAR EVLDSRGNPT VEVEVLTESG AFGRALVPSG ASTGEHEAVE LRDGDKSRYL GKGVTKAVEN VNEIIAPEII EGEFSVLDQV SIDKMMIALD GTPNKGKLGA NAILGVSIAV ARAAADLLGQ PLYKYLGGFN GKQLPVPMMN IVNGGSHSDA PIAFQEFMIL PVGATTFKES LRWGTEIFHN LKSILSKRGL ETAVGDEGGF APKFEGTEDA VETIIQAIEA AGYKPGEEVF LGFDCASSEF YENGVYDYSK FEGEHGAKRT AAEQVDYLEQ LVDKYPIITI EDGMDENDWD GWKQLTERIG DRVQLVGDDL FVTNTEILAK GIENGIGNSI LIKVNQIGTL TETFDAIEMA QKAGYTAVVS HRSGETEDTT IADIAVATNA GQIKTGSLSR TDRIAKYNQL LRIEDELFET AKYDGIKSFY NLDK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.