missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Type II iodothyronine deiodinase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Type II iodothyronine deiodinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9482-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_113908.3 | |
Type II iodothyronine deiodinase Protein | |
Research Use Only | |
Dio2 | |
Recombinant Protein | |
Yeast | |
MGLLSVDLLI TLQILPVFFS NCLFLALYDS VILLKHVALL LSRSKSTRGE WRRMLTSEGL RCVWNSFLLD AYKQVKLGED APNSSVVHVS NPEAGNNCAS EKTADGAECH LLDFASAERP LVVNFGSATU PPFTRQLPAF RQLVEEFSSV ADFLLVYIDE AHPSDGWAVP GDSSMSFEVK KHRNQEDRCA AAHQLLERFS LPPQCQVVAD RMDNNANVAY GVAFERVCIV QRRKIAYLGG KGPFSYNLQE VRSWLEKNFS KRUILD | |
Greater than 90% as determined by SDS-PAGE. |
65162 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |