missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Type II iodothyronine deiodinase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Type II iodothyronine deiodinase, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_113908.3 |
---|---|
Gene ID (Entrez) | 65162 |
Name | Type II iodothyronine deiodinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098401
|
enQuireBio™
QP9482-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16038411
|
enQuireBio™
QP9482-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16008411
|
enQuireBio™
QP9482-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16028411
|
enQuireBio™
QP9482-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048411
|
enQuireBio™
QP9482-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16018411
|
enQuireBio™
QP9482-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_113908.3 | |
Type II iodothyronine deiodinase Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
65162 | |
Research Use Only | |
Dio2 | |
Recombinant Protein | |
Yeast | |
MGLLSVDLLI TLQILPVFFS NCLFLALYDS VILLKHVALL LSRSKSTRGE WRRMLTSEGL RCVWNSFLLD AYKQVKLGED APNSSVVHVS NPEAGNNCAS EKTADGAECH LLDFASAERP LVVNFGSATU PPFTRQLPAF RQLVEEFSSV ADFLLVYIDE AHPSDGWAVP GDSSMSFEVK KHRNQEDRCA AAHQLLERFS LPPQCQVVAD RMDNNANVAY GVAFERVCIV QRRKIAYLGG KGPFSYNLQE VRSWLEKNFS KRUILD | |
Greater than 90% as determined by SDS-PAGE. |