missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TLR4 / CD284 Protein
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
Specifications
Specifications
| Accession Number | NP_062051.1 |
| Gene ID (Entrez) | 29260 |
| Name | TLR4 / CD284 Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | Tlr4 |
| Protein Length | Extracellular Domain |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rat |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction