missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Semaphorin 3A / SEMA3A Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Semaphorin 3A / SEMA3A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7692-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070221

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_059006.1
Semaphorin 3A / SEMA3A Protein
Research Use Only
Sema3a
Recombinant Protein
E. coli
YANGKNNVPR LKLSYKEMLE SNNVITFNGL ANSSSYHTFL LDEERSRLYV GAKDHIFSFN LVNIKDFQKI VWPVSYTRRD ECKWAGKDIL KECANFIKVL KAYNQTHLYA CGTGAFHPIC TYIEVGHHPE DNIFKLQDSH FENGRGKSPY DPKLLTASLL IDGELYSGTA ADFMGRDFAI FRTLGHHHPI RTEQHDSRWL NDPRFISAHL IPESDNPEDD KVYFFFRENA IDGEHSGKAT HARIGQICKN DFGGHRSLVN KWTTFLKARL ICSVPGPNGI DTHFDELQDV FLMNSKDPKN PIVYGVFTTS SNIFKGSAVC MYSMSDVRRV FLGPYAHRDG PNYQWVPYQG RVPYPRPGTC PSKTFGGFDS TKDLPDDVIT FARSHPAMYN PVFPINNRPI MIKTDVNYQF TQIVVDRVDA EDGQYDVMFI GTDVGTVLKV VSVPKETWHD LEEVLLEEMT VFREPTTISA MELSTKQQQL YIGSTAGVAQ LPLHRCDIYG KACAECCLAR DPYCAWDGSS CSRYFPTAKR RTRRQDIRNG DPLTHCSDLQ HHDNHHGHSL EERIIYGVEN SSTFLECSPK SQRALVYWQF QRRNEDRKEE IRVGDHIIRT EQGLLLRSLQ KKDSGNYLCH AVEHGFMQTL LKVTLEVIDT EHLEELLHKD DDGDGSKTKE MSSSMTPSQK VWYRDFMQLI NHPNLNTMDE FCEQVWKRDR KQRRQRPGHS QGSSNKWKHM QESKKGRNRR THEFERAPRS V
Greater than 90% as determined by SDS-PAGE.
29751
50 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers