missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Semaphorin 3A / SEMA3A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Semaphorin 3A / SEMA3A, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_059006.1 |
---|---|
Gene ID (Entrez) | 29751 |
Name | Semaphorin 3A / SEMA3A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16020221
|
enQuireBio™
QP7692-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16040221
|
enQuireBio™
QP7692-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16030221
|
enQuireBio™
QP7692-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16050221
|
enQuireBio™
QP7692-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16060221
|
enQuireBio™
QP7692-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070221
|
enQuireBio™
QP7692-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_059006.1 | |
Semaphorin 3A / SEMA3A Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
29751 | |
Research Use Only | |
Sema3a | |
Recombinant Protein | |
E. coli | |
YANGKNNVPR LKLSYKEMLE SNNVITFNGL ANSSSYHTFL LDEERSRLYV GAKDHIFSFN LVNIKDFQKI VWPVSYTRRD ECKWAGKDIL KECANFIKVL KAYNQTHLYA CGTGAFHPIC TYIEVGHHPE DNIFKLQDSH FENGRGKSPY DPKLLTASLL IDGELYSGTA ADFMGRDFAI FRTLGHHHPI RTEQHDSRWL NDPRFISAHL IPESDNPEDD KVYFFFRENA IDGEHSGKAT HARIGQICKN DFGGHRSLVN KWTTFLKARL ICSVPGPNGI DTHFDELQDV FLMNSKDPKN PIVYGVFTTS SNIFKGSAVC MYSMSDVRRV FLGPYAHRDG PNYQWVPYQG RVPYPRPGTC PSKTFGGFDS TKDLPDDVIT FARSHPAMYN PVFPINNRPI MIKTDVNYQF TQIVVDRVDA EDGQYDVMFI GTDVGTVLKV VSVPKETWHD LEEVLLEEMT VFREPTTISA MELSTKQQQL YIGSTAGVAQ LPLHRCDIYG KACAECCLAR DPYCAWDGSS CSRYFPTAKR RTRRQDIRNG DPLTHCSDLQ HHDNHHGHSL EERIIYGVEN SSTFLECSPK SQRALVYWQF QRRNEDRKEE IRVGDHIIRT EQGLLLRSLQ KKDSGNYLCH AVEHGFMQTL LKVTLEVIDT EHLEELLHKD DDGDGSKTKE MSSSMTPSQK VWYRDFMQLI NHPNLNTMDE FCEQVWKRDR KQRRQRPGHS QGSSNKWKHM QESKKGRNRR THEFERAPRS V | |
Greater than 90% as determined by SDS-PAGE. |