missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDGF-BB Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 2.0 ng/mL, corresponding to a specific activity of > 5.0 x 10^5 IU/mg.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 1 x PBS, pH 6.9. |
| Gene ID (Entrez) | 5155 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 24.4 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of PDGF-BB as determined by LAL method. |
| Gene Alias | c-sis;IBGC5;PDGF2;PDGF-2;PDGFB;Platelet-derived growth factor subunit B;SIS;SSV |
| Gene Symbol | PDGFB |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?