Learn More
enQuireBio™ Recombinant Rat PCSK1 / NEC1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PCSK1 / NEC1, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_058787.1 |
---|---|
Gene ID (Entrez) | 25204 |
Name | PCSK1 / NEC1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038101
|
enQuireBio™
QP9322-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16078101
|
enQuireBio™
QP9322-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16048101
|
enQuireBio™
QP9322-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16068101
|
enQuireBio™
QP9322-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16088101
|
enQuireBio™
QP9322-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058101
|
enQuireBio™
QP9322-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_058787.1 | |
PCSK1 / NEC1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25204 | |
Research Use Only | |
Pcsk1 | |
Recombinant Protein | |
Yeast | |
SVPRDSALNL FNDPMWNQQW YLQDTRMTAS LPKLDLHVIP VWQKGITGKG VVITVLDDGL EWNHTDIYAN YDPEASYDFN DNDHDPFPRY DPTNENKHGT RCAGEIAMQA NNHKCGVGVA YNSKVGGIRM LDGIVTDAIE ASSIGFNPGH VDIYSASWGP NDDGKTVEGP GRLAQKAFEY GVKQGRQGKG SIFVWASGNG GRQGDNCDCD GYTDSIYTIS ISSASQQGLS PWYAEKCSST LATSYSSGDY TDQRITSADL HNDCTETHTG TSASAPLAAG IFALALEANP NLTWRDMQHL VVWTSEYDPL ANNPGWKKNG AGLMVNSRFG FGLLNAKALV DLADPRTWRN VPEKKECIIK DNNFEPRALK ANGEVIVEIP TRACEGQENA INSLEHVQFE ATIEYSRRGD LHVTLTSAAG TSTVLLAERE RDTSPNGFKN WDFMSVHTWG ENPVGTWTLK VTDMSGRMQN EGRIVNWKLI LHGTSSQPEH MKQPRVYTSY NTVQNDRRGV EKMVNVVEEK PTQNSLNGNL LVPKNSSSSS VEDRRDEQVQ GAPSKAMLRL LQSAFSKNTP SKQSSKIPSA KLSVPYEGLY EALEKLNKPS QLEDSEDSLY SDYVDVFYNT KPYKHRDDRL LQALMDILNE KN | |
Greater than 90% as determined by SDS-PAGE. |